Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069374.10 | NP_501775.1 | 7 - 8 | 1159 | 67 | |
GenBank mRNA | AF083223.1 | - | 6 - 7 | 1076 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |