Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dur-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027974.4 | NP_001023145.1 | 8 - 9 | 521 | 62 | |
NM_001268504.3 | NP_001255433.1 | 11 - 12 | 716 | 62 | ||
NM_001268505.3 | NP_001255434.1 | 12 - 13 | 782 | 62 | ||
NM_001268506.3 | NP_001255435.1 | 12 - 13 | 785 | 62 | ||
NM_001268507.3 | NP_001255436.1 | 14 - 15 | 1619 | 62 | ||
NM_001268510.3 | NP_001255439.1 | 5 - 6 | 299 | 62 | ||
NM_001268514.3 | NP_001255443.1 | 1 - 2 | 56 | 62 | ||
NM_001307013.3 | NP_001293942.1 | 8 - 9 | 452 | 62 | ||
NM_001372910.1 | NP_001360167.1 | 8 - 9 | 521 | 62 | ||
NM_001380423.1 | NP_001368122.1 | 1 - 2 | 56 | 62 | ||
NM_001383174.1 | 8 - 9 | 522 | 62 | |||
NM_001383175.1 | 8 - 9 | 521 | 62 | |||
NM_069385.6 | NP_501786.2 | 12 - 13 | 1497 | 62 | ||
NM_069386.8 | NP_501787.3 | 13 - 14 | 1550 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |