Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dur-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027974.4 | NP_001023145.1 | 4 - 5 | 244 | 83 | |
NM_001268504.3 | NP_001255433.1 | 7 - 8 | 439 | 83 | ||
NM_001268505.3 | NP_001255434.1 | 8 - 9 | 505 | 83 | ||
NM_001268506.3 | NP_001255435.1 | 8 - 9 | 508 | 83 | ||
NM_001268507.3 | NP_001255436.1 | 10 - 11 | 1342 | 83 | ||
NM_001307013.3 | NP_001293942.1 | 4 - 5 | 175 | 83 | ||
NM_001372910.1 | NP_001360167.1 | 4 - 5 | 244 | 83 | ||
NM_001383173.1 | 4 - 5 | 249 | 83 | |||
NM_001383174.1 | 4 - 5 | 245 | 83 | |||
NM_001383175.1 | 4 - 5 | 244 | 83 | |||
NM_069385.6 | NP_501786.2 | 8 - 9 | 1220 | 83 | ||
NM_069386.8 | NP_501787.3 | 9 - 10 | 1273 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |