Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268528.3 | NP_001255457.1 | 2 - 3 | 478 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |