Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028122.5 | NP_001023293.2 | 4 - 5 | 361 | 81 | |
NM_001028127.6 | NP_001023298.1 | 14 - 15 | 1536 | 81 | ||
NM_001028133.5 | NP_001023304.1 | 4 - 5 | 846 | 81 | ||
NM_001129316.4 | NP_001122788.1 | 4 - 5 | 489 | 81 | ||
NM_001129317.5 | NP_001122789.1 | 3 - 4 | 678 | 81 | ||
NM_001136391.4 | NP_001129863.1 | 4 - 5 | 319 | 81 | ||
NM_001268551.3 | NP_001255480.1 | 16 - 17 | 1714 | 81 | ||
NM_001268552.3 | NP_001255481.1 | 5 - 6 | 469 | 81 | ||
NM_001268553.3 | NP_001255482.1 | 4 - 5 | 565 | 81 | ||
NM_001372918.2 | NP_001359633.1 | 15 - 16 | 1661 | 81 | ||
NM_001380439.2 | NP_001366993.1 | 15 - 16 | 1682 | 81 | ||
NM_001380440.2 | NP_001366994.1 | 16 - 17 | 1789 | 81 | ||
NM_001380441.2 | NP_001366996.1 | 10 - 11 | 1237 | 81 | ||
NM_001380442.1 | NP_001366992.1 | 11 - 12 | 3804 | 81 | ||
NM_001380443.1 | NP_001366997.1 | 10 - 11 | 1068 | 81 | ||
NM_001383186.1 | 13 - 14 | 1428 | 81 | |||
NM_001383187.1 | 14 - 15 | 1543 | 81 | |||
GenBank mRNA | AF180735.1 | - | 12 - 13 | 1330 | 81 | |
AF233262.1 | - | 12 - 13 | 1231 | 81 | ||
AF233263.1 | - | 12 - 13 | 1270 | 81 | ||
AF233264.1 | - | 13 - 14 | 1384 | 81 | ||
AF233265.1 | - | 12 - 13 | 1225 | 81 | ||
AF233266.1 | - | 10 - 11 | 1083 | 81 | ||
AF233267.1 | - | 10 - 11 | 1068 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |