Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cal-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268562.3 | NP_001255491.1 | 1 - 2 | 301 | 150 | |
NM_001383190.1 | 5 - 6 | 708 | 150 | |||
NM_001383191.1 | 4 - 5 | 561 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |