Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fmo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069567.7 | NP_501968.2 | 4 - 5 | 674 | 107 | |
GenBank mRNA | AJ582070.1 | - | 3 - 4 | 563 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |