Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdz-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069617.4 | NP_502018.2 | 5 - 6 | 552 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |