Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069710.6 | NP_502111.3 | 22 - 23 | 3597 | 156 | |
GenBank mRNA | AY726541.1 | - | 22 - 23 | 3527 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |