Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047531.2 | NP_001040996.1 | 10 - 11 | 1329 | 75 | |
NM_001047532.6 | NP_001040997.1 | 9 - 10 | 1127 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |