Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307532.3 | NP_001294461.1 | 2 - 3 | 412 | 119 | |
NM_067599.4 | NP_500000.3 | 2 - 3 | 412 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |