Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307625.3 | NP_001294554.1 | 14 - 15 | 3005 | 97 | |
NM_001307626.3 | NP_001294555.1 | 11 - 12 | 2438 | 97 | ||
NM_001307627.3 | NP_001294556.1 | 11 - 12 | 2438 | 97 | ||
NM_001307628.3 | NP_001294557.1 | 2 - 3 | 692 | 97 | ||
NM_001307629.3 | NP_001294558.1 | 2 - 3 | 692 | 97 | ||
NM_067617.6 | NP_500018.3 | 14 - 15 | 3005 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |