Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdt-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380036.2 | NP_001367680.1 | 2 - 3 | 323 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |