Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307551.3 | NP_001294480.1 | 1 - 2 | 51 | 68 | |
NM_067672.5 | NP_500073.3 | 3 - 4 | 329 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |