Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380115.1 | NP_001368197.1 | 1 - 2 | 143 | 93 | |
NM_001383060.1 | 10 - 11 | 2413 | 93 |
Gene Symbol: Y54G2A.40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356912.3 | NP_001343694.1 | 2544 | 93 |
Gene Symbol: clec-84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380113.2 | NP_001368086.1 | 11 - 12 | 4839 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |