Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307526.4 | NP_001294455.1 | 2 - 3 | 195 | 78 | |
NM_067995.4 | NP_500396.2 | 2 - 3 | 199 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |