Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068030.2 | NP_500431.1 | 4 - 5 | 589 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |