Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068103.5 | NP_500504.1 | 3 - 4 | 495 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |