Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307636.3 | NP_001294565.1 | 1 - 2 | 408 | 60 | |
NM_068104.6 | NP_500505.1 | 2 - 3 | 501 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |