Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkh-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027976.6 | NP_001023147.1 | 1 - 2 | 513 | 64 | |
NM_001306882.4 | NP_001293811.1 | 1 - 2 | 498 | 64 | ||
NM_001306884.4 | NP_001293813.1 | 1 - 2 | 498 | 64 | ||
NM_001306885.3 | NP_001293814.1 | 1 - 2 | 498 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |