Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306838.3 | NP_001293767.1 | 1 - 2 | 228 | 85 | |
NM_068493.8 | NP_500894.1 | 3 - 4 | 959 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |