Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028211.4 | NP_001023382.1 | 11 - 12 | 1524 | 84 | |
NR_131543.1 | 11 - 12 | 1533 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |