Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smg-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380273.1 | NP_001368064.1 | 3 - 4 | 656 | 65 | |
NM_068573.9 | NP_500974.2 | 3 - 4 | 660 | 65 | ||
GenBank mRNA | EF472000.1 | - | 3 - 4 | 674 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |