Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307154.4 | NP_001294083.1 | 7 - 8 | 2143 | 110 | |
NM_069848.7 | NP_502249.3 | 7 - 8 | 2140 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |