Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gex-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069967.6 | NP_502368.1 | 1 - 2 | 117 | 93 | |
GenBank mRNA | AB073210.1 | - | 1 - 2 | 113 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |