Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: him-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069989.7 | NP_502390.2 | 1 - 2 | 307 | 77 | |
GenBank mRNA | AY095296.1 | - | 1 - 2 | 329 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |