Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dod-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268760.2 | NP_001255689.1 | 3 - 4 | 469 | 76 | |
NM_001268761.3 | NP_001255690.1 | 2 - 3 | 266 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |