Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msh-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070130.8 | NP_502531.1 | 3 - 4 | 391 | 94 | |
GenBank mRNA | AF271389.1 | - | 3 - 4 | 391 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |