Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mau-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268805.3 | NP_001255734.1 | 3 - 4 | 595 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |