Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047487.3 | NP_001040952.2 | 6 - 7 | 967 | 105 | |
NM_001047488.4 | NP_001040953.1 | 3 - 4 | 397 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |