Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noah-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070298.6 | NP_502699.1 | 1 - 2 | 153 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |