Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tra-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070350.6 | NP_502751.1 | 8 - 9 | 1859 | 81 | |
NR_131640.1 | 7 - 8 | 2032 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |