Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307187.2 | NP_001294116.1 | 4 - 5 | 596 | 73 | |
NM_001307188.3 | NP_001294117.1 | 4 - 5 | 569 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |