Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269027.3 | NP_001255956.2 | 13 - 14 | 1731 | 103 | |
NM_001269028.2 | NP_001255957.2 | 12 - 13 | 1761 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |