Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380283.2 | NP_001368373.1 | 13 - 14 | 2147 | 95 | |
NM_068617.8 | NP_501018.1 | 18 - 19 | 2926 | 95 | ||
GenBank mRNA | AF480439.1 | - | 18 - 19 | 2921 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |