Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyci-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372852.3 | NP_001360494.1 | 8 | 1396 | 84 | |
NM_001383110.1 | 7 - 8 | 1309 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |