Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sru-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313162.1 | NP_001300091.1 | 2 - 3 | 237 | 135 | |
NM_070888.2 | NP_503289.2 | 2 - 3 | 432 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |