Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313356.3 | NP_001300285.1 | 2 - 3 | 427 | 88 | |
NM_001313357.3 | NP_001300286.1 | 1 - 2 | 156 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Caenorhabditis elegans Probe spans exons |