Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071079.4 | NP_503480.3 | 3 - 4 | 578 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |