Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313328.1 | NP_001300257.1 | 4 - 5 | 619 | 114 | |
NM_071086.1 | NP_503487.1 | 5 - 6 | 892 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |