Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313323.4 | NP_001300252.1 | 2 - 3 | 1100 | 70 | |
NM_001313324.3 | NP_001300253.1 | 1 - 2 | 413 | 70 | ||
NM_071111.6 | NP_503512.1 | 4 - 5 | 1262 | 70 | ||
GenBank mRNA | AF031841.1 | - | 4 - 5 | 1269 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |