Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071184.3 | NP_503585.3 | 3 - 4 | 513 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |