Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038398.3 | NP_001033487.1 | 3 - 4 | 587 | 177 | |
NM_001316801.3 | NP_001303730.1 | 1 - 2 | 126 | 177 | ||
NR_133578.1 | 3 - 4 | 456 | 177 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |