Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071210.5 | NP_503611.3 | 4 - 5 | 655 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |