Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071649.2 | NP_504050.1 | 2 - 3 | 136 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |