Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-35A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071723.2 | NP_504124.1 | 6 - 7 | 1372 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |