Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071854.4 | NP_504255.3 | 4 - 5 | 470 | 135 | |
NR_136380.1 | 6 - 7 | 757 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |