Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nex-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322553.3 | NP_001309625.1 | 5 - 6 | 584 | 74 | |
NM_001380638.1 | NP_001368593.1 | 5 - 6 | 626 | 74 | ||
NM_001383302.1 | 5 - 6 | 618 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |