Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071923.4 | NP_504324.1 | 2 - 3 | 1179 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |