Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071985.6 | NP_504386.2 | 6 - 7 | 847 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |